siRNA Details
Gene Symbol: Efcab3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001081046.1 | NP_001074515.1 | 9 | 1177 | ||
GenBank mRNA | AK014861.1 | - | 9 | 1172 | ||
BC050801.1 | - | 9 | 1175 | 6770929 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Efcab3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001081046.1 | NP_001074515.1 | 9 | 1177 | ||
GenBank mRNA | AK014861.1 | - | 9 | 1172 | ||
BC050801.1 | - | 9 | 1175 | 6770929 |