siRNA Details
Gene Symbol: Fam71d | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_027597.4 | NP_081873.2 | 3, 4 | 486 | ||
NM_029069.1 | NP_083345.1 | 3, 4 | 491 | |||
GenBank mRNA | AB473726.1 | - | 1, 2 | 166 | ||
AK014851.1 | - | 3, 4 | 492 | |||
AK015802.1 | - | 3, 4 | 339 | |||
BC051149.1 | - | 3, 4 | 346 | 6707726 |
