siRNA Details
| Gene Symbol: 4921517D22Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_183290.2 | NP_899113.1 | 4 | 798 | ||
| GenBank mRNA | AK014909.1 | - | 4 | 799 | ||
| AK133255.1 | - | 4 | 806 | |||
| BC147600.1 | - | 3 | 587 | 9056590 | ||
| BC147624.1 | - | 3 | 587 | 9056614 | ||

4921517D22Rik
Genomic Map