siRNA Details
| Gene Symbol: Trim69 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_080510.2 | NP_536771.2 | 2 | 637 | ||
| GenBank mRNA | AF334958.1 | - | 2 | 637 | ||
| AK014923.1 | - | 2 | 631 | |||
| BC050815.1 | - | 2 | 629 | 6774293 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Trim69| Gene Symbol: Trim69 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_080510.2 | NP_536771.2 | 2 | 637 | ||
| GenBank mRNA | AF334958.1 | - | 2 | 637 | ||
| AK014923.1 | - | 2 | 631 | |||
| BC050815.1 | - | 2 | 629 | 6774293 | ||