siRNA Details
| Gene Symbol: Nol8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001271397.1 | NP_001258326.1 | 7 | 1450 | ||
| GenBank mRNA | AK014994.1 | - | 7 | 1451 | ||
| AK029961.1 | - | 7 | 1163 | |||
| AK147172.1 | - | 7 | 1186 | |||
| BC049166.1 | - | 7 | 1041 | |||
| BC058699.1 | - | 7 | 1606 | |||
| BC125575.1 | - | 7 | 1176 | 40130090 | ||
| BC125579.1 | - | 7 | 1176 | 40130094 | ||

Nol8
Genomic Map