siRNA Details
| Gene Symbol: Pgm2l1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_027629.3 | NP_081905.1 | 6 | 1018 | ||
| GenBank mRNA | AK016437.1 | - | 4 | 377 | ||
| AK039189.1 | - | 6 | 1002 | |||
| AK040513.1 | - | 6 | 938 | |||
| BC020351.1 | - | 6 | 1231 | |||
| BC032150.1 | - | 2 | 224 | 5004308 | ||
| BC076571.1 | - | 6 | 915 | 30356163 | ||

Pgm2l1
Genomic Map