siRNA Details
| Gene Symbol: 4931429I11Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001081121.1 | NP_001074590.1 | 6 | 1893 | ||
| GenBank mRNA | AK016487.1 | - | 2 | 759 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

4931429I11Rik| Gene Symbol: 4931429I11Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001081121.1 | NP_001074590.1 | 6 | 1893 | ||
| GenBank mRNA | AK016487.1 | - | 2 | 759 | ||