siRNA Details
| Gene Symbol: Phf6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001290546.1 | NP_001277475.1 | 3, 4 | 398 | ||
| NM_027642.2 | NP_081918.1 | 5, 6 | 684 | |||
| GenBank mRNA | AK016480.1 | - | 5, 6 | 685 | ||
| AK028498.1 | - | 3, 4 | 320 | |||
| AK038320.1 | - | 5, 6 | 604 | |||
| AK122560.1 | - | 5, 6 | 614 | |||
| AK141804.1 | - | 5, 6 | 627 | |||
| AK163345.1 | - | 5, 6 | 640 | |||
| BC043127.1 | - | 5, 6 | 632 | 6410361 | ||
| BC055330.1 | - | 5, 6 | 627 | 6406483 | ||
| BC057374.1 | - | 5, 6 | 590 | 6826091 | ||

Phf6
Genomic Map