siRNA Details
| Gene Symbol: D7Ertd443e | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001081331.1 | NP_001074800.1 | 3 | 1481 | ||
| NM_001199941.1 | NP_001186870.1 | 4 | 1615 | |||
| GenBank mRNA | AK016590.1 | - | 3 | 376 | ||
| AK145411.1 | - | 3 | 1482 | |||
| GQ499374.1 | - | 4 | 1615 | |||

D7Ertd443e
Genomic Map