siRNA Details
Gene Symbol: D7Ertd443e | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001081331.1 | NP_001074800.1 | 5 | 1712 | ||
NM_001199941.1 | NP_001186870.1 | 6 | 1846 | |||
GenBank mRNA | AK016590.1 | - | 5 | 607 | ||
AK145411.1 | - | 5 | 1713 | |||
GQ499374.1 | - | 6 | 1846 |
