siRNA Details
| Gene Symbol: Uvssa | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001081101.2 | NP_001074570.1 | 4, 5 | 849 | ||
| GenBank mRNA | AK016722.1 | - | 3, 4 | 510 | ||
| AK028507.1 | - | 3, 4 | 1218 | |||
| AK040506.1 | - | 4, 5 | 853 | |||
| AK129384.1 | - | 4, 5 | 535 | |||
| AK156117.1 | - | 3, 4 | 1008 | |||
| BC137803.1 | - | 4, 5 | 773 | 8860822 | ||
| BC144920.1 | - | 4, 5 | 688 | 9053468 | ||

Uvssa
Genomic Map