siRNA Details
| Gene Symbol: Uvssa | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001081101.2 | NP_001074570.1 | 7 | 1452 | ||
| GenBank mRNA | AK016722.1 | - | 6 | 1113 | ||
| AK028507.1 | - | 6 | 1821 | |||
| AK040506.1 | - | 7 | 1456 | |||
| AK129384.1 | - | 7 | 1138 | |||
| AK156117.1 | - | 6 | 1611 | |||
| BC061483.1 | - | 2 | 207 | |||
| BC137803.1 | - | 7 | 1376 | 8860822 | ||
| BC144920.1 | - | 7 | 1291 | 9053468 | ||

Uvssa
Genomic Map