siRNA Details
Gene Symbol: Zdhhc11 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_027704.2 | NP_081980.1 | 4 | 533 | ||
GenBank mRNA | AK016865.1 | - | 4 | 544 | ||
AY668948.1 | - | 3 | 474 | |||
BC116803.1 | - | 4 | 533 | 40126122 | ||
BC116807.1 | - | 4 | 533 | 40126126 | ||
BC144831.1 | - | 4 | 525 | |||
BC144832.1 | - | 4 | 533 |
