siRNA Details
| Gene Symbol: Nbas | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_027706.1 | NP_081982.1 | 34, 35 | 4104 | ||
| GenBank mRNA | BC046406.1 | - | 17, 18 | 2137 | ||
| BC057020.1 | - | 10, 11 | 1197 | 6822193 | ||
| BC072598.1 | - | 20, 21 | 2610 | |||

Nbas
Genomic Map