siRNA Details
| Gene Symbol: Nbas | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_027706.1 | NP_081982.1 | 35 | 4134 | ||
| GenBank mRNA | BC046406.1 | - | 18 | 2167 | ||
| BC057020.1 | - | 11 | 1227 | 6822193 | ||
| BC072598.1 | - | 21 | 2640 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Nbas| Gene Symbol: Nbas | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_027706.1 | NP_081982.1 | 35 | 4134 | ||
| GenBank mRNA | BC046406.1 | - | 18 | 2167 | ||
| BC057020.1 | - | 11 | 1227 | 6822193 | ||
| BC072598.1 | - | 21 | 2640 | |||