siRNA Details
| Gene Symbol: Nipbl | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_027707.3 | NP_081983.2 | 45, 46 | 8257 | ||
| NM_201232.2 | NP_957684.1 | 45, 46 | 8257 | |||
| GenBank mRNA | AJ627033.1 | - | 45, 46 | 8257 | ||
| AJ640138.1 | - | 45, 46 | 8257 | |||
| AK014915.1 | - | 1, 2 | 119 | |||
| BC049151.1 | - | 4, 5 | 781 | |||
| BC055787.1 | - | 1, 2 | 68 | |||

Nipbl
Genomic Map