siRNA Details
| Gene Symbol: Cage1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_027724.2 | NP_082000.2 | 9 | 2411 | ||
| GenBank mRNA | AK132733.1 | - | 5 | 959 | ||
| AY695827.1 | - | 8 | 2146 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Cage1| Gene Symbol: Cage1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_027724.2 | NP_082000.2 | 9 | 2411 | ||
| GenBank mRNA | AK132733.1 | - | 5 | 959 | ||
| AY695827.1 | - | 8 | 2146 | |||