siRNA Details
Gene Symbol: Ccdc57 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_027745.1 | NP_082021.1 | 19 | 3029 | ||
GenBank mRNA | AK017054.1 | - | 4 | 496 | ||
BC030385.1 | - | 6 | 816 | |||
BC056480.1 | - | 19 | 3029 | 5689542 |
