siRNA Details
| Gene Symbol: Slc29a3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_023596.3 | NP_076085.1 | 6 | 1404 | ||
| GenBank mRNA | AF326986.1 | - | 6 | 1415 | ||
| AK034133.1 | - | 6 | 1400 | |||
| AK051317.1 | - | 6 | 1429 | |||
| AK157868.1 | - | 5 | 1242 | |||
| BC137864.1 | - | 6 | 1387 | 8860883 | ||
| BC144983.1 | - | 6 | 1388 | |||

Slc29a3
Genomic Map