siRNA Details
| Gene Symbol: Arhgap26 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_175164.4 | NP_780373.3 | 1 | 439 | ||
| GenBank mRNA | AK011368.1 | - | 1 | 442 | ||
| AK129176.1 | - | 1 | 377 | |||
| BC141394.1 | - | 1 | 308 | 9056007 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Arhgap26| Gene Symbol: Arhgap26 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_175164.4 | NP_780373.3 | 1 | 439 | ||
| GenBank mRNA | AK011368.1 | - | 1 | 442 | ||
| AK129176.1 | - | 1 | 377 | |||
| BC141394.1 | - | 1 | 308 | 9056007 | ||