siRNA Details
| Gene Symbol: Tchhl1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_027762.3 | NP_082038.2 | 3 | 1475 | ||
| GenBank mRNA | AK017244.1 | - | 3 | 1476 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Tchhl1| Gene Symbol: Tchhl1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_027762.3 | NP_082038.2 | 3 | 1475 | ||
| GenBank mRNA | AK017244.1 | - | 3 | 1476 | ||