siRNA Details
| Gene Symbol: Riok1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_024242.3 | NP_077204.2 | 16 | 1927 | ||
| GenBank mRNA | AK014182.1 | - | 3 | 654 | ||
| AK017312.1 | - | 16 | 1622 | |||
| AK152707.1 | - | 16 | 1676 | |||
| AK161315.1 | - | 17 | 2170 | |||
| BC002158.1 | - | 10 | 884 | 3485522 | ||

Riok1
Genomic Map