siRNA Details
Gene Symbol: Wdr31 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001290521.1 | NP_001277450.1 | 3, 4 | 434 | ||
NM_023597.2 | NP_076086.1 | 3, 4 | 327 | |||
GenBank mRNA | AF274321.1 | - | 3, 4 | 304 | ||
AK141026.1 | - | 3, 4 | 432 | |||
BC026500.1 | - | 2, 3 | 213 | |||
BC037102.1 | - | 3, 4 | 317 | 5364307 | ||
CK618879.1 | - | 3, 4 | 434 |
