siRNA Details
| Gene Symbol: Prr16 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001081224.2 | NP_001074693.1 | 2 | 214 | ||
| GenBank mRNA | AK017282.1 | - | 1 | 28 | ||
| AK042678.1 | - | 375 | ||||
| AK045646.1 | - | 1 | 168 | |||
| AK046456.1 | - | 2 | 379 | |||
| AK081911.1 | - | 190 | ||||
| BC132481.1 | - | 2 | 214 | 40130758 | ||

Prr16
Genomic Map