siRNA Details
Gene Symbol: Foxn3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_183186.2 | NP_899009.2 | 5, 6 | 894 | ||
GenBank mRNA | AK017346.1 | - | 4, 5 | 697 | ||
AK046748.1 | - | 5, 6 | 1169 | |||
BC099971.1 | - | 5, 6 | 972 | 30008715 |
