siRNA Details
| Gene Symbol: Pex1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001293806.1 | NP_001280735.1 | 19, 20 | 3359 | ||
| NM_027777.2 | NP_082053.1 | 18, 19 | 3239 | |||
| GenBank mRNA | AK017309.1 | - | 6, 7 | 656 | ||
| BC090845.1 | - | 18, 19 | 3239 | 6400629 | ||

Pex1
Genomic Map