siRNA Details
Gene Symbol: Chd6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | BC011423.1 | - | 1 | 53 | ||
BC049798.1 | - | 5 | 1035 | |||
BC072648.1 | - | 6 | 1695 | |||
BC145629.1 | - | 35 | 7006 | |||
BC150806.1 | - | 36 | 7210 | 9087717 | ||
CN664285.1 | - | 3 | 486 |
