siRNA Details
| Gene Symbol: Chd6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK086807.1 | - | 2 | 129 | ||
| BC011423.1 | - | 2 | 224 | |||
| BC049798.1 | - | 6 | 1206 | |||
| BC072648.1 | - | 7 | 1866 | |||
| BC145629.1 | - | 36 | 7177 | |||
| BC150806.1 | - | 37 | 7381 | 9087717 | ||

Chd6
Genomic Map