siRNA Details
| Gene Symbol: Chd6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | BC027287.1 | - | 2 | 411 | ||
| BC049798.1 | - | 3 | 515 | |||
| BC072648.1 | - | 4 | 1176 | |||
| BC145629.1 | - | 33 | 6486 | |||
| BC150806.1 | - | 34 | 6690 | 9087717 | ||
| CN526286.1 | - | 4 | 576 | |||
| CO433513.1 | - | 4 | 549 | |||

Chd6
Genomic Map