siRNA Details
| Gene Symbol: Kctd6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK017384.1 | - | 3 | 437 | ||
| AK082924.1 | - | 3 | 436 | |||
| AK159067.1 | - | 3 | 1849 | |||
| BB625950.1 | - | 3 | 326 | |||
| BC096370.1 | - | 3 | 276 | 3597940 | ||
| BC131971.1 | - | 2 | 246 | 40130248 | ||
| BC131975.1 | - | 2 | 246 | 40130252 | ||
| CX239507.1 | - | 3 | 503 | |||

Kctd6
Genomic Map