siRNA Details
Gene Symbol: Fmnl2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_172409.2 | NP_765997.2 | 16 | 2162 | ||
GenBank mRNA | AF513716.1 | - | 5 | 559 | ||
BC048004.1 | - | 1 | 50 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Fmnl2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_172409.2 | NP_765997.2 | 16 | 2162 | ||
GenBank mRNA | AF513716.1 | - | 5 | 559 | ||
BC048004.1 | - | 1 | 50 |