siRNA Details
| Gene Symbol: Usp19 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001168371.2 | NP_001161843.1 | 24 | 3747 | ||
| NM_001168372.2 | NP_001161844.1 | 24 | 3744 | |||
| NM_001168373.2 | NP_001161845.1 | 24 | 3741 | |||
| NM_027804.4 | NP_082080.3 | 24 | 3744 | |||
| NM_145407.3 | NP_663382.2 | 23 | 3672 | |||
| GenBank mRNA | AK043088.1 | - | 23 | 3508 | ||
| AK122396.2 | - | 23 | 3530 | |||
| AK144099.1 | - | 19 | 2775 | |||
| AK146504.1 | - | 24 | 3570 | |||
| AK148412.1 | - | 19 | 2732 | |||
| AK159473.1 | - | 24 | 3566 | |||
| AK160807.1 | - | 24 | 3570 | |||
| AK171942.1 | - | 24 | 3573 | |||
| AK180108.1 | - | 2 | 193 | |||
| BC006916.1 | - | 16 | 2278 | |||
| BC046824.1 | - | 15 | 2207 | |||
| BC060613.1 | - | 24 | 3672 | 6400986 | ||

Usp19
Genomic Map