siRNA Details
| Gene Symbol: Bbs7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_027810.3 | NP_082086.2 | 19 | 2222 | ||
| GenBank mRNA | AF521645.1 | - | 19 | 2221 | ||
| AK132661.1 | - | 19 | 2150 | |||
| BC031505.1 | - | 19 | 2172 | 3494587 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Bbs7| Gene Symbol: Bbs7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_027810.3 | NP_082086.2 | 19 | 2222 | ||
| GenBank mRNA | AF521645.1 | - | 19 | 2221 | ||
| AK132661.1 | - | 19 | 2150 | |||
| BC031505.1 | - | 19 | 2172 | 3494587 | ||