siRNA Details
| Gene Symbol: Afmid | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_027827.3 | NP_082103.1 | 3 | 263 | ||
| GenBank mRNA | AF399717.1 | - | 3 | 258 | ||
| AK018563.1 | - | 1 | 199 | |||
| AY099479.1 | - | 3 | 258 | |||
| BC024452.1 | - | 3 | 234 | |||
| BC043309.1 | - | 3 | 229 | |||
| BC066780.1 | - | 3 | 242 | 30301832 | ||
| BY780917.1 | - | 3 | 263 | |||

Afmid
Genomic Map