siRNA Details
| Gene Symbol: Myo1e | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_181072.3 | NP_851417.2 | 8 | 1044 | ||
| GenBank mRNA | AK029145.1 | - | 8 | 1046 | ||
| BC040786.1 | - | 9 | 1100 | |||
| BC051391.1 | - | 8 | 1026 | 5053090 | ||
| CX242330.1 | - | 3 | 242 | |||

Myo1e
Genomic Map