siRNA Details
Gene Symbol: Myo1e | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_181072.3 | NP_851417.2 | 11 | 1489 | ||
GenBank mRNA | AK029145.1 | - | 11 | 1491 | ||
BB220783.2 | - | 1 | 132 | |||
BC040786.1 | - | 12 | 1545 | |||
BC051391.1 | - | 11 | 1471 | 5053090 | ||
CX242330.1 | - | 6 | 687 |
