siRNA Details
| Gene Symbol: Tmem248 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001081394.1 | NP_001074863.1 | 5 | 899 | ||
| NM_027854.1 | NP_082130.1 | 5 | 961 | |||
| GenBank mRNA | AK002297.1 | - | 5 | 899 | ||
| AK142176.1 | - | 5 | 949 | |||
| AK171152.1 | - | 5 | 927 | |||
| BC033455.1 | - | 5 | 871 | 4018584 | ||

Tmem248
Genomic Map