siRNA Details
| Gene Symbol: Acy3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AA110692.1 | - | 1 | 30 | ||
| AF356878.1 | - | 8 | 1056 | |||
| AF356879.1 | - | 7 | 915 | |||
| AF375479.1 | - | 6 | 867 | |||
| AK002247.1 | - | 8 | 1055 | |||
| AK143782.1 | - | 8 | 1058 | |||
| AY169234.1 | - | 8 | 1055 | |||
| BC010795.1 | - | 6 | 1276 | 4221711 | ||

Acy3
Genomic Map