siRNA Details
| Gene Symbol: Wdr27 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_175173.3 | NP_780382.2 | 21 | 2142 | ||
| GenBank mRNA | AK040117.1 | - | 6 | 582 | ||
| AK077050.1 | - | 21 | 2143 | |||
| AV277048.2 | - | 1 | 60 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Wdr27| Gene Symbol: Wdr27 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_175173.3 | NP_780382.2 | 21 | 2142 | ||
| GenBank mRNA | AK040117.1 | - | 6 | 582 | ||
| AK077050.1 | - | 21 | 2143 | |||
| AV277048.2 | - | 1 | 60 | |||