siRNA Details
| Gene Symbol: Wdr27 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_175173.3 | NP_780382.2 | 3 | 239 | ||
| GenBank mRNA | AK077050.1 | - | 3 | 240 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Wdr27| Gene Symbol: Wdr27 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_175173.3 | NP_780382.2 | 3 | 239 | ||
| GenBank mRNA | AK077050.1 | - | 3 | 240 | ||