siRNA Details
| Gene Symbol: Galnt14 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_027864.2 | NP_082140.2 | 15 | 1941 | ||
| GenBank mRNA | AB175681.1 | - | 15 | 1549 | ||
| AK049598.1 | - | 15 | 1887 | |||
| AK078292.1 | - | 15 | 1554 | |||
| BC037716.1 | - | 9 | 920 | |||
| BC117800.1 | - | 15 | 1725 | 40092267 | ||

Galnt14
Genomic Map