siRNA Details
| Gene Symbol: Galnt14 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_027864.2 | NP_082140.2 | 8, 9 | 1218 | ||
| GenBank mRNA | AB175681.1 | - | 8, 9 | 826 | ||
| AK002747.1 | - | 8, 9 | 1190 | |||
| AK049598.1 | - | 8, 9 | 1164 | |||
| AK078292.1 | - | 8, 9 | 831 | |||
| BC037716.1 | - | 2, 3 | 197 | |||
| BC117800.1 | - | 8, 9 | 1002 | 40092267 | ||

Galnt14
Genomic Map