siRNA Details
| Gene Symbol: Galnt14 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_027864.2 | NP_082140.2 | 2 | 603 | ||
| GenBank mRNA | AB175681.1 | - | 2 | 211 | ||
| AK002747.1 | - | 2 | 575 | |||
| AK049598.1 | - | 2 | 549 | |||
| AK078292.1 | - | 2 | 216 | |||
| AK090146.1 | - | 2 | 606 | |||
| BC117800.1 | - | 2 | 387 | 40092267 | ||

Galnt14
Genomic Map