siRNA Details
| Gene Symbol: Arhgef3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001289686.1 | NP_001276615.1 | 10, 11 | 1125 | ||
| NM_001289687.1 | NP_001276616.1 | 7, 8 | 996 | |||
| NM_001289688.1 | NP_001276617.1 | 7, 8 | 1170 | |||
| NM_027871.2 | NP_082147.1 | 8, 9 | 970 | |||
| GenBank mRNA | AK004600.2 | - | 8, 9 | 941 | ||
| AK029872.1 | - | 11, 12 | 1155 | |||
| BC005517.1 | - | 7, 8 | 967 | 3494309 | ||
| BC007153.1 | - | 1, 2 | 214 | |||
| BC012262.1 | - | 7, 8 | 948 | 4192378 | ||

Arhgef3
Genomic Map