siRNA Details
| Gene Symbol: Arhgef3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001289686.1 | NP_001276615.1 | 13 | 1595 | ||
| NM_001289687.1 | NP_001276616.1 | 10 | 1466 | |||
| NM_001289688.1 | NP_001276617.1 | 10 | 1640 | |||
| NM_027871.2 | NP_082147.1 | 11 | 1440 | |||
| GenBank mRNA | AK004600.2 | - | 11 | 1411 | ||
| AK029872.1 | - | 14 | 1625 | |||
| BC005517.1 | - | 10 | 1437 | 3494309 | ||
| BC007153.1 | - | 4 | 684 | |||
| BC012262.1 | - | 10 | 1418 | 4192378 | ||

Arhgef3
Genomic Map