siRNA Details
Gene Symbol: Telo2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001163661.1 | NP_001157133.1 | 2 | 643 | ||
NM_027880.2 | NP_082156.2 | 2 | 554 | |||
GenBank mRNA | AK004582.1 | - | 2 | 424 | ||
AK028658.1 | - | 2 | 408 | |||
AK050773.1 | - | 2 | 374 | |||
AK084830.1 | - | 2 | 424 | |||
AK129189.1 | - | 2 | 592 | |||
AK141388.1 | - | 2 | 620 | |||
AK143827.1 | - | 2 | 644 | |||
AK146963.1 | - | 2 | 430 | |||
AK154727.1 | - | 2 | 456 | |||
BC011077.1 | - | 2 | 404 | 3984911 | ||
BC013127.1 | - | 2 | 630 | |||
BC080713.1 | - | 2 | 583 |
