siRNA Details
| Gene Symbol: Osbpl3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001163645.1 | NP_001157117.1 | 21, 22 | 2955 | ||
| NM_027881.3 | NP_082157.2 | 21, 22 | 2673 | |||
| GenBank mRNA | AK004768.1 | - | 21, 22 | 2675 | ||
| AK042104.1 | - | 1, 2 | 1052 | |||
| BC044866.1 | - | 20, 21 | 2600 | 5003631 | ||
| BC086485.1 | - | 21, 22 | 2867 | 5344992 | ||
| CF751761.1 | - | 2, 3 | 222 | |||

Osbpl3
Genomic Map