siRNA Details
| Gene Symbol: Osbpl3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001163645.1 | NP_001157117.1 | 5, 6 | 845 | ||
| NM_027881.3 | NP_082157.2 | 4, 5 | 455 | |||
| GenBank mRNA | AK004768.1 | - | 4, 5 | 457 | ||
| AK037114.1 | - | 4, 5 | 455 | |||
| AK160310.1 | - | 5, 6 | 846 | |||
| AK173006.1 | - | 5, 6 | 654 | |||
| BC044866.1 | - | 4, 5 | 490 | 5003631 | ||
| BC086485.1 | - | 5, 6 | 757 | 5344992 | ||

Osbpl3
Genomic Map