siRNA Details
| Gene Symbol: Fam13c | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001143776.1 | NP_001137248.1 | 12 | 1526 | ||
| NM_001143777.1 | NP_001137249.1 | 13 | 1633 | |||
| NM_024244.4 | NP_077206.3 | 12 | 1529 | |||
| GenBank mRNA | AK004797.1 | - | 12 | 1513 | ||
| AK032053.1 | - | 14 | 1368 | |||
| AK041147.1 | - | 12 | 1527 | |||
| AK220421.1 | - | 12 | 3071 | |||
| BC003231.1 | - | 1 | 28 | |||
| BC129839.1 | - | 12 | 1482 | 40060810 | ||
| BC129840.1 | - | 12 | 1485 | 40060811 | ||

Fam13c
Genomic Map