siRNA Details
| Gene Symbol: Vps11 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_027889.1 | NP_082165.1 | 10 | 1663 | ||
| GenBank mRNA | AK004695.1 | - | 10 | 1638 | ||
| AK032484.1 | - | 10 | 1628 | |||
| AK037658.1 | - | 9 | 2014 | |||
| AK090030.1 | - | 10 | 1644 | |||
| AK153963.1 | - | 10 | 1636 | |||
| AK162897.1 | - | 10 | 1638 | |||
| AK180848.1 | - | 2 | 145 | |||
| BC029004.1 | - | 10 | 1663 | 5360111 | ||

Vps11
Genomic Map